Assay Details
| Gene Symbol: LOC100775304 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255498.2 | NP_001242427.1 | 2 - 3 | 213 | 127 | |
| GenBank mRNA | AK244400.1 | - | 2 - 3 | 272 | 127 | |
| BT096163.1 | - | 2 - 3 | 216 | 127 | ||
More Information
Set Membership: |
|

LOC100775304
Genomic Map