Assay Details
| Gene Symbol: LOC100812311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252821.1 | NP_001239750.1 | 9 | 916 | 118 | |
| GenBank mRNA | BT097872.1 | - | 9 | 916 | 118 | |
More Information
Set Membership: |
|

LOC100812311
Genomic Map