Assay Details
| Gene Symbol: LOC100806459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253025.2 | NP_001239954.1 | 2 | 918 | 93 | |
| GenBank mRNA | BT095819.1 | - | 2 | 920 | 93 | |
More Information
Set Membership: |
|

LOC100806459
Genomic Map