Assay Details
| Gene Symbol: LOC100815008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254175.1 | NP_001241104.1 | 4 | 518 | 132 | |
| GenBank mRNA | AB550120.1 | - | 4 | 518 | 132 | |
| FJ573245.1 | - | 4 | 480 | 132 | ||
More Information
Set Membership: |
|

LOC100815008
Genomic Map