Assay Details
| Gene Symbol: PHT1-14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254235.1 | NP_001241164.1 | 2 | 1522 | 91 | |
| GenBank mRNA | FJ797414.1 | - | 2 | 1522 | 91 | |
More Information
Set Membership: |
|

PHT1-14
Genomic Map