Assay Details
Gene Symbol: LOC100785835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001254242.1 | NP_001241171.1 | 6 | 916 | 138 | |
GenBank mRNA | BT093287.1 | - | 6 | 916 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
