Assay Details
| Gene Symbol: LOC100790724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254391.2 | NP_001241320.1 | 3 | 1022 | 134 | |
| GenBank mRNA | BT097734.1 | - | 3 | 1024 | 134 | |
More Information
Set Membership: |
|

LOC100790724
Genomic Map