Assay Details
| Gene Symbol: LOC100785104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT098903.1 | - | 7 | 860 | 181 | |
| L14930.1 | - | 7 | 967 | 181 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100785104| Gene Symbol: LOC100785104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT098903.1 | - | 7 | 860 | 181 | |
| L14930.1 | - | 7 | 967 | 181 | ||
Set Membership: |
|