Assay Details
| Gene Symbol: LOC100793323 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254604.2 | NP_001241533.1 | 5 | 1119 | 139 | |
| GenBank mRNA | BT097811.1 | - | 5 | 1121 | 139 | |
More Information
Set Membership: |
|

LOC100793323
Genomic Map