Assay Details
| Gene Symbol: LOC100803065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254670.1 | NP_001241599.1 | 1 | 111 | 79 | |
| GenBank mRNA | AK244410.1 | - | 1 | 191 | 79 | |
| BT095794.1 | - | 1 | 111 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT095794.1, NM_001254670.1 transcripts.
Set Membership: |
|

LOC100803065
Genomic Map