Assay Details
| Gene Symbol: LOC100812565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255031.1 | NP_001241960.1 | 9 | 1327 | 192 | |
| GenBank mRNA | BT094270.1 | - | 9 | 1327 | 192 | |
More Information
Set Membership: |
|

LOC100812565
Genomic Map