Assay Details
| Gene Symbol: LOC100808983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255037.1 | NP_001241966.1 | 7 | 870 | 106 | |
| GenBank mRNA | BT096684.1 | - | 7 | 870 | 106 | |
More Information
Set Membership: |
|

LOC100808983
Genomic Map