Assay Details
| Gene Symbol: LOC100779919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255530.1 | NP_001242459.1 | 3 | 812 | 127 | |
| GenBank mRNA | BT094587.1 | - | 3 | 812 | 127 | |
More Information
Set Membership: |
|

LOC100779919
Genomic Map