Assay Details
| Gene Symbol: LOC100805078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255813.1 | NP_001242742.1 | 1 | 77 | 115 | |
| GenBank mRNA | BT093617.1 | - | 1 | 77 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT093617.1, NM_001255813.1 transcripts.
Set Membership: |
|

LOC100805078
Genomic Map