Assay Details
| Gene Symbol: NLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289275.1 | NP_001276204.1 | 3 | 1018 | 115 | |
| GenBank mRNA | JX392398.1 | - | 3 | 1018 | 115 | |
More Information
Set Membership: |
|

NLP1
Genomic Map