Assay Details
| Gene Symbol: GME1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289329.1 | NP_001276258.1 | 6 | 1047 | 87 | |
| GenBank mRNA | FJ619111.1 | - | 6 | 1047 | 87 | |
| Gene Symbol: LOC100797618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287273.1 | - | 7 | 1112 | 87 | |
More Information
Set Membership: |
|

GME1
Genomic Map