Assay Details
| Gene Symbol: LOC100816354 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289363.1 | NP_001276292.1 | 1 | 123 | 108 | |
| GenBank mRNA | AK244405.1 | - | 2 | 108 | ||
| AK286689.1 | - | 2 | 108 | |||
| JX644158.1 | - | 1 | 123 | 108 | ||
More Information
Set Membership: |
|

LOC100816354
Genomic Map