Assay Details
| Gene Symbol: MYB54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248694.2 | NP_001235623.1 | 2 | 713 | 78 | |
| GenBank mRNA | DQ822881.1 | - | 2 | 714 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB54| Gene Symbol: MYB54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248694.2 | NP_001235623.1 | 2 | 713 | 78 | |
| GenBank mRNA | DQ822881.1 | - | 2 | 714 | 78 | |
Set Membership: |
|