Assay Details
| Gene Symbol: ERF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244940.1 | - | 2 | 1122 | 121 | |
| BT096814.1 | - | 2 | 1111 | 121 | ||
| JN416602.1 | - | 2 | 960 | 121 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERF7| Gene Symbol: ERF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK244940.1 | - | 2 | 1122 | 121 | |
| BT096814.1 | - | 2 | 1111 | 121 | ||
| JN416602.1 | - | 2 | 960 | 121 | ||
Set Membership: |
|