Assay Details
| Gene Symbol: LOC100786069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254114.1 | NP_001241043.1 | 6 | 1093 | 128 | |
| GenBank mRNA | BT096732.1 | - | 6 | 1093 | 128 | |
More Information
Set Membership: |
|

LOC100786069
Genomic Map