Assay Details
| Gene Symbol: LOC100786397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254527.1 | NP_001241456.1 | 1 | 420 | 77 | |
| GenBank mRNA | AK245682.1 | - | 1 | 359 | 77 | |
| BT093650.1 | - | 1 | 420 | 77 | ||
More Information
Set Membership: |
|

LOC100786397
Genomic Map