Assay Details
| Gene Symbol: LOC100791834 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255049.2 | NP_001241978.1 | 6 | 1318 | 109 | |
| GenBank mRNA | BT094333.1 | - | 6 | 1319 | 109 | |
More Information
Set Membership: |
|

LOC100791834
Genomic Map