Assay Details
| Gene Symbol: LOC100818762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255250.1 | NP_001242179.1 | 1 | 121 | 62 | |
| GenBank mRNA | BT092661.1 | - | 1 | 121 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BT092661.1, NM_001255250.1 transcripts.
Set Membership: |
|

LOC100818762
Genomic Map