Assay Details
| Gene Symbol: LOC100778363 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255438.2 | NP_001242367.1 | 3 | 1166 | 82 | |
| GenBank mRNA | AK285954.1 | - | 3 | 1167 | 82 | |
| BT098915.1 | - | 3 | 1168 | 82 | ||
More Information
Set Membership: |
|

LOC100778363
Genomic Map