Assay Details
| Gene Symbol: LOC100778108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255441.1 | NP_001242370.1 | 5 | 1590 | 97 | |
| GenBank mRNA | AK285395.1 | - | 5 | 1611 | 97 | |
| BT096947.1 | - | 5 | 1590 | 97 | ||
More Information
Set Membership: |
|

LOC100778108
Genomic Map