Assay Details
| Gene Symbol: LOC100783260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001255585.2 | NP_001242514.1 | 3 | 1020 | 72 | |
| GenBank mRNA | AK243909.1 | - | 3 | 983 | 72 | |
| BT097437.1 | - | 3 | 1022 | 72 | ||
More Information
Set Membership: |
|

LOC100783260
Genomic Map