Assay Details
| Gene Symbol: LOC547791 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251039.1 | NP_001237968.1 | 4 | 1714 | 110 | |
| GenBank mRNA | AK287379.1 | - | 3 | 1677 | 110 | |
| L08632.1 | - | 4 | 1714 | 110 | ||
More Information
Set Membership: |
|

LOC547791
Genomic Map