Assay Details
| Gene Symbol: Ybx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871824.2 | XP_004871881.1 | 3 - 4 | 418 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ybx1| Gene Symbol: Ybx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871824.2 | XP_004871881.1 | 3 - 4 | 418 | 87 | |
Set Membership: |
|