Assay Details
| Gene Symbol: Dpagt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856769.3 | XP_004856826.1 | 2 - 3 | 459 | 61 | |
| XM_004856770.3 | XP_004856827.1 | 2 - 3 | 459 | 61 | ||
| XM_013076539.2 | XP_012931993.1 | 3 - 4 | 493 | 61 | ||
| XM_013076541.2 | XP_012931995.1 | 3 - 4 | 522 | 61 | ||
| XM_021237658.1 | XP_021093317.1 | 2 - 3 | 459 | 61 | ||
More Information
Set Membership: |
|

Dpagt1
Genomic Map