Assay Details
| Gene Symbol: LOC101715399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838650.2 | XP_004838707.1 | 2 - 3 | 413 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101715399| Gene Symbol: LOC101715399 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838650.2 | XP_004838707.1 | 2 - 3 | 413 | 99 | |
Set Membership: |
|