Assay Details
| Gene Symbol: Nipsnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843407.2 | XP_004843464.1 | 7 - 8 | 665 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nipsnap1| Gene Symbol: Nipsnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843407.2 | XP_004843464.1 | 7 - 8 | 665 | 66 | |
Set Membership: |
|