Assay Details
Gene Symbol: Arcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004856716.2 | XP_004856773.1 | 3 - 4 | 611 | 98 | |
XM_021237733.1 | XP_021093392.1 | 4 - 5 | 656 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
