Assay Details
| Gene Symbol: Polr3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848294.2 | XP_004848351.1 | 2 - 3 | 247 | 96 | |
| XM_004848296.2 | XP_004848353.1 | 2 - 3 | 330 | 96 | ||
| XM_004848297.3 | XP_004848354.1 | 2 - 3 | 246 | 96 | ||
| XM_021261238.1 | XP_021116897.1 | 2 - 3 | 329 | 96 | ||
More Information
Set Membership: |
|

Polr3d
Genomic Map