Assay Details
| Gene Symbol: Tomm40l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858655.3 | XP_004858712.1 | 5 - 6 | 670 | 62 | |
| XM_021239676.1 | XP_021095335.1 | 5 - 6 | 556 | 62 | ||
More Information
Set Membership: |
|

Tomm40l
Genomic Map