Assay Details
| Gene Symbol: Matr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841827.3 | XP_004841884.1 | 2 - 3 | 1249 | 84 | |
| XM_004841828.2 | XP_004841885.1 | 5 - 6 | 1449 | 84 | ||
| XM_021255901.1 | XP_021111560.1 | 5 - 6 | 1450 | 84 | ||
| XM_021255902.1 | XP_021111561.1 | 2 - 3 | 1115 | 84 | ||
| XM_021255903.1 | XP_021111562.1 | 2 - 3 | 1168 | 84 | ||
| XM_021255904.1 | XP_021111563.1 | 2 - 3 | 1112 | 84 | ||
| XM_021255905.1 | XP_021111564.1 | 5 - 6 | 1450 | 84 | ||
| XM_021255906.1 | XP_021111565.1 | 5 - 6 | 1450 | 84 | ||
More Information
Set Membership: |
|

Matr3
Genomic Map