Assay Details
| Gene Symbol: Tfap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004847707.3 | XP_004847764.1 | 6 - 7 | 1244 | 73 | |
| XM_004847708.3 | XP_004847765.1 | 6 - 7 | 1161 | 73 | ||
| XM_004847709.3 | XP_004847766.1 | 6 - 7 | 1851 | 73 | ||
More Information
Set Membership: |
|

Tfap2a
Genomic Map