Assay Details
| Gene Symbol: Etv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871264.2 | XP_004871321.1 | 2 - 3 | 352 | 66 | |
| XM_004871265.2 | XP_004871322.1 | 2 - 3 | 356 | 66 | ||
More Information
Set Membership: |
|

Etv3
Genomic Map