Assay Details
| Gene Symbol: LOC101700724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861275.3 | XP_004861332.1 | 2 - 3 | 1058 | 94 | |
| XM_013078312.2 | XP_012933766.1 | 2 - 3 | 171 | 94 | ||
More Information
Set Membership: |
|

LOC101700724
Genomic Map