Assay Details
| Gene Symbol: Ppp1cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843809.2 | XP_004843866.1 | 5 - 6 | 971 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1cc| Gene Symbol: Ppp1cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843809.2 | XP_004843866.1 | 5 - 6 | 971 | 102 | |
Set Membership: |
|