Assay Details
| Gene Symbol: Ddx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870239.3 | XP_004870296.1 | 11 - 12 | 1469 | 99 | |
| XM_004870241.3 | XP_004870298.1 | 11 - 12 | 1304 | 99 | ||
More Information
Set Membership: |
|

Ddx5
Genomic Map