Assay Details
| Gene Symbol: Uprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867490.3 | XP_004867547.1 | 3 - 4 | 537 | 80 | |
| XM_004867491.3 | XP_004867548.1 | 3 - 4 | 527 | 80 | ||
More Information
Set Membership: |
|

Uprt
Genomic Map