Assay Details
| Gene Symbol: Lsm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855367.3 | XP_004855424.1 | 2 - 3 | 438 | 97 | |
| XM_004855368.3 | XP_004855425.1 | 2 - 3 | 312 | 97 | ||
| XM_021236812.1 | XP_021092471.1 | 3 - 4 | 381 | 97 | ||
More Information
Set Membership: |
|

Lsm1
Genomic Map