Assay Details
| Gene Symbol: Egr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021239145.1 | XP_021094804.1 | 2 - 3 | 326 | 71 | |
| XM_021239146.1 | XP_021094805.1 | 2 - 3 | 689 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021239146.1 transcripts.
Set Membership: |
|

Egr2
Genomic Map