Assay Details
| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921160.2 | XP_004921217.1 | 2 - 3 | 319 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Suv39h2| Gene Symbol: Suv39h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004921160.2 | XP_004921217.1 | 2 - 3 | 319 | 99 | |
Set Membership: |
|