Assay Details
| Gene Symbol: LOC101708403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845613.3 | XP_004845670.1 | 6 - 7 | 874 | 62 | |
| XM_004845614.3 | XP_004845671.1 | 6 - 7 | 877 | 62 | ||
| XM_013072565.2 | XP_012928019.1 | 6 - 7 | 877 | 62 | ||
| XM_013072566.2 | XP_012928020.1 | 6 - 7 | 874 | 62 | ||
| XM_021258995.1 | XP_021114654.1 | 6 - 7 | 843 | 62 | ||
| XM_021258996.1 | XP_021114655.1 | 6 - 7 | 877 | 62 | ||
More Information
Set Membership: |
|

LOC101708403
Genomic Map