Assay Details
| Gene Symbol: Nr1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859457.2 | XP_004859514.1 | 4 - 5 | 1213 | 66 | |
| XM_021240368.1 | XP_021096027.1 | 5 - 6 | 2134 | 66 | ||
| XM_021240369.1 | XP_021096028.1 | 4 - 5 | 597 | 66 | ||
More Information
Set Membership: |
|

Nr1d1
Genomic Map