Assay Details
| Gene Symbol: Wdr20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836926.3 | XP_004836983.1 | 1 - 2 | 310 | 103 | |
| XM_004836928.3 | XP_004836985.1 | 1 - 2 | 309 | 103 | ||
| XM_004836932.3 | XP_004836989.1 | 1 - 2 | 310 | 103 | ||
| XM_021244037.1 | XP_021099696.1 | 1 - 2 | 312 | 103 | ||
More Information
Set Membership: |
|

Wdr20
Genomic Map