Assay Details
| Gene Symbol: Rps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285851.1 | NP_001272780.1 | 1 - 2 | 139 | 56 | |
| XM_013066870.2 | XP_012922324.1 | 2 - 3 | 341 | 56 | ||
| XM_013066871.2 | XP_012922325.1 | 2 - 3 | 270 | 56 | ||
More Information
Set Membership: |
|

Rps7
Genomic Map