Assay Details
| Gene Symbol: Aifm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310225.1 | NP_001297154.1 | 8 - 9 | 1102 | 57 | |
| XM_004846191.3 | XP_004846248.1 | 8 - 9 | 1127 | 57 | ||
| GenBank mRNA | KM486684.1 | - | 8 - 9 | 1102 | 57 | |
More Information
Set Membership: |
|

Aifm2
Genomic Map