Assay Details
| Gene Symbol: Krt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310297.1 | NP_001297226.1 | 4 - 5 | 779 | 100 | |
| GenBank mRNA | KM486731.1 | - | 4 - 5 | 779 | 100 | |
More Information
Set Membership: |
|

Krt8
Genomic Map